Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604910 MIM: 606419 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNOT3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CNOT3 - CCR4-NOT transcription complex subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014516.3 | Intron | NP_055331.1 | ||||
XM_005278279.1 | Intron | XP_005278336.1 | ||||
XM_005278280.2 | Intron | XP_005278337.1 | ||||
XM_005278281.1 | Intron | XP_005278338.1 | ||||
XM_005278282.2 | Intron | XP_005278339.1 | ||||
XM_011526992.1 | Intron | XP_011525294.1 | ||||
XM_011526993.2 | Intron | XP_011525295.1 |
PRPF31 - pre-mRNA processing factor 31 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |