Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616804 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
VSTM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
VSTM1 - V-set and transmembrane domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288791.1 | 1232 | UTR 3 | NP_001275720.1 | |||
NM_001288792.1 | 1232 | UTR 3 | NP_001275721.1 | |||
NM_001288793.1 | 1232 | UTR 3 | NP_001275722.1 | |||
NM_198481.3 | 1232 | UTR 3 | NP_940883.2 | |||
XM_011526845.1 | 1232 | UTR 3 | XP_011525147.1 | |||
XM_011526846.1 | 1232 | UTR 3 | XP_011525148.1 | |||
XM_011526847.1 | 1232 | UTR 3 | XP_011525149.1 | |||
XM_011526848.1 | 1232 | UTR 3 | XP_011525150.1 | |||
XM_011526849.1 | 1232 | UTR 3 | XP_011525151.1 | |||
XM_017026666.1 | 1232 | Intron | XP_016882155.1 |
Set Membership: |
HapMap Validated |