Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGTATCTAAGGGCACCAAGAAGA[C/T]GCGATTAGACGTGTAAGCGTGACTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605611 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SH3BP4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SH3BP4 - SH3 domain binding protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014521.2 | 1658 | Intron | NP_055336.1 | |||
XM_011510891.2 | 1658 | Intron | XP_011509193.1 | |||
XM_011510892.1 | 1658 | Intron | XP_011509194.1 | |||
XM_011510893.1 | 1658 | Intron | XP_011509195.1 | |||
XM_011510894.2 | 1658 | UTR 5 | XP_011509196.1 |