Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGGAATGTGATCCCCAGGGGGCCG[A/G]GGAGGGCCCTGGGGAGGAGCCTGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJC22 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNAJC22 - DnaJ heat shock protein family (Hsp40) member C22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304944.1 | 926 | UTR 5 | NP_001291873.1 | |||
NM_024902.3 | 926 | Intron | NP_079178.2 | |||
XM_005269155.2 | 926 | Intron | XP_005269212.1 | |||
XM_005269156.2 | 926 | Intron | XP_005269213.1 | |||
XM_005269157.2 | 926 | Intron | XP_005269214.1 |