Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTATTCCTTATATATGAGATAACA[A/G]TAACATCCTCCACAGGGTGCCATGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TUBA1C PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TUBA1C - tubulin alpha 1c | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303114.1 | Intron | NP_001290043.1 | ||||
NM_001303115.1 | Intron | NP_001290044.1 | ||||
NM_001303116.1 | Intron | NP_001290045.1 | ||||
NM_001303117.1 | Intron | NP_001290046.1 | ||||
NM_032704.4 | Intron | NP_116093.1 |
Set Membership: |
HapMap |