Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCCGGGCTGCGGGGCGCAGCATTG[C/T]GCGGTCATGGTGGGCCAGATGTACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLBD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PLBD2 - phospholipase B domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159727.1 | 29 | UTR 5 | NP_001153199.1 | |||
NM_173542.3 | 29 | UTR 5 | NP_775813.2 | |||
XM_011538023.2 | 29 | Intron | XP_011536325.1 | |||
XM_017018977.1 | 29 | UTR 5 | XP_016874466.1 |