Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142703 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HRH2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HRH2 - histamine receptor H2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001131055.1 | Intron | NP_001124527.1 | ||||
NM_022304.2 | Intron | NP_071640.1 | ||||
XM_005265904.3 | Intron | XP_005265961.1 | ||||
XM_006714864.3 | Intron | XP_006714927.1 | ||||
XM_006714865.3 | Intron | XP_006714928.1 | ||||
XM_011534548.2 | Intron | XP_011532850.1 | ||||
XM_011534549.2 | Intron | XP_011532851.1 | ||||
XM_017009426.1 | Intron | XP_016864915.1 |