Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164013 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NFRKB PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NFRKB - nuclear factor related to kappaB binding protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRDM10 - PR domain 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020228.2 | 6188 | UTR 3 | NP_064613.2 | |||
NM_199437.1 | 6188 | UTR 3 | NP_955469.1 | |||
NM_199438.1 | 6188 | UTR 3 | NP_955470.1 | |||
NM_199439.1 | 6188 | UTR 3 | NP_955471.1 | |||
XM_011542912.2 | 6188 | UTR 3 | XP_011541214.1 | |||
XM_017018023.1 | 6188 | UTR 3 | XP_016873512.1 |
Set Membership: |
HapMap |