Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610916 MIM: 184753 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NSUN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NSUN2 - NOP2/Sun RNA methyltransferase family member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SRD5A1 - steroid 5 alpha-reductase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001047.3 | Intron | NP_001038.1 | ||||
NM_001324322.1 | Intron | NP_001311251.1 | ||||
NM_001324323.1 | Intron | NP_001311252.1 |
Set Membership: |
HapMap |