Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACACCAGACTTTCTGTGTAAAAAC[C/T]GACTGTAGATAACCTGACCTATCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612471 MIM: 176761 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AGXT2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
AGXT2 - alanine--glyoxylate aminotransferase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRLR - prolactin receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000949.6 | Intron | NP_000940.1 | ||||
NM_001204314.2 | Intron | NP_001191243.1 | ||||
NM_001204315.1 | Intron | NP_001191244.1 | ||||
NM_001204316.1 | Intron | NP_001191245.1 | ||||
NM_001204317.1 | Intron | NP_001191246.1 | ||||
NM_001204318.1 | Intron | NP_001191247.1 | ||||
XM_006714484.2 | Intron | XP_006714547.1 | ||||
XM_011514068.2 | Intron | XP_011512370.1 | ||||
XM_011514069.2 | Intron | XP_011512371.1 | ||||
XM_017009645.1 | Intron | XP_016865134.1 | ||||
XM_017009646.1 | Intron | XP_016865135.1 |
Set Membership: |
HapMap |