Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATGGAGTGGGTGGGGCGGCTCGG[A/G]CTACAGTTCAATCTCGAATGTCTTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604018 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUMBL PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NUMBL - NUMB like, endocytic adaptor protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289979.1 | 1921 | UTR 3 | NP_001276908.1 | |||
NM_001289980.1 | 1921 | UTR 3 | NP_001276909.1 | |||
NM_004756.4 | 1921 | UTR 3 | NP_004747.1 |
Set Membership: |
HapMap |