Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGTCTAGGTCATTGAAAAATCCAA[C/T]GAAAGCTACAAACACATTTTTCAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616384 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
UGT3A2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
UGT3A2 - UDP glycosyltransferase family 3 member A2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168316.1 | Intron | NP_001161788.1 | ||||
NM_174914.3 | Intron | NP_777574.2 | ||||
XM_011513988.2 | Intron | XP_011512290.1 | ||||
XM_017009150.1 | Intron | XP_016864639.1 |