Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATCTGTCATTCTGTGCAAGTGTGT[A/G]TCCCATTGATTTGAGTATGTTGAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF525 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF525 - zinc finger protein 525 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF845 - zinc finger protein 845 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321522.1 | 5495 | UTR 3 | NP_001308451.1 | |||
NM_001321523.1 | 5495 | UTR 3 | NP_001308452.1 | |||
NM_001321524.1 | 5495 | UTR 3 | NP_001308453.1 | |||
NM_138374.2 | 5495 | UTR 3 | NP_612383.1 |