Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604820 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LILRB3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LILRB3 - leukocyte immunoglobulin like receptor B3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001081450.2 | Intron | NP_001074919.2 | ||||
NM_001320960.1 | Intron | NP_001307889.1 | ||||
NM_006864.3 | Intron | NP_006855.3 | ||||
XM_011526381.2 | Intron | XP_011524683.1 | ||||
XM_011526382.2 | Intron | XP_011524684.1 | ||||
XM_017026218.1 | Intron | XP_016881707.1 | ||||
XM_017026219.1 | Intron | XP_016881708.1 | ||||
XM_017026220.1 | Intron | XP_016881709.1 | ||||
XM_017026221.1 | Intron | XP_016881710.1 | ||||
XM_017026222.1 | Intron | XP_016881711.1 | ||||
XM_017026223.1 | Intron | XP_016881712.1 |
Set Membership: |
HapMap |