Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608746 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC25A23 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLC25A23 - solute carrier family 25 member 23 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024103.2 | Intron | NP_077008.2 | ||||
XM_011528274.1 | Intron | XP_011526576.1 | ||||
XM_011528275.1 | Intron | XP_011526577.1 | ||||
XM_011528276.1 | Intron | XP_011526578.1 | ||||
XM_011528277.1 | Intron | XP_011526579.1 | ||||
XM_011528278.1 | Intron | XP_011526580.1 | ||||
XM_011528279.1 | Intron | XP_011526581.1 | ||||
XM_011528280.1 | Intron | XP_011526582.1 | ||||
XM_011528281.1 | Intron | XP_011526583.1 | ||||
XM_011528282.1 | Intron | XP_011526584.1 | ||||
XM_011528283.1 | Intron | XP_011526585.1 | ||||
XM_011528284.1 | Intron | XP_011526586.1 | ||||
XM_011528285.1 | Intron | XP_011526587.1 | ||||
XM_017027285.1 | Intron | XP_016882774.1 | ||||
XM_017027286.1 | Intron | XP_016882775.1 | ||||
XM_017027287.1 | Intron | XP_016882776.1 | ||||
XM_017027288.1 | Intron | XP_016882777.1 | ||||
XM_017027289.1 | Intron | XP_016882778.1 | ||||
XM_017027290.1 | Intron | XP_016882779.1 | ||||
XM_017027291.1 | Intron | XP_016882780.1 | ||||
XM_017027292.1 | Intron | XP_016882781.1 |
Set Membership: |
HapMap |