Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAACAGCCCAGGACCCCAGCCTCC[C/T]GCTTCCTTCTTTCCTGTGGCTCCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603842 MIM: 610057 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NDUFB7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NDUFB7 - NADH:ubiquinone oxidoreductase subunit B7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004146.5 | Intron | NP_004137.2 | ||||
XM_011528039.2 | Intron | XP_011526341.1 |
TECR - trans-2,3-enoyl-CoA reductase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |