Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGGCATCTGTGAAATCTTTTAAGC[A/G]TAGCTCTGTTTCCAAACTGACTTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611595 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TXNL4A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TXNL4A - thioredoxin like 4A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303471.2 | Intron | NP_001290400.1 | ||||
NM_001305557.1 | Intron | NP_001292486.1 | ||||
NM_001305563.1 | Intron | NP_001292492.1 | ||||
NM_001305564.1 | Intron | NP_001292493.1 | ||||
NM_006701.4 | Intron | NP_006692.1 |
Set Membership: |
HapMap |