Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602204 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BICD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BICD1 - BICD cargo adaptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003398.1 | Intron | NP_001003398.1 | ||||
NM_001714.2 | Intron | NP_001705.2 | ||||
XM_005253465.3 | Intron | XP_005253522.1 | ||||
XM_005253466.2 | Intron | XP_005253523.1 | ||||
XM_006719134.3 | Intron | XP_006719197.1 | ||||
XM_011520812.2 | Intron | XP_011519114.1 | ||||
XM_011520813.1 | Intron | XP_011519115.1 | ||||
XM_011520814.2 | Intron | XP_011519116.1 | ||||
XM_011520815.2 | Intron | XP_011519117.1 | ||||
XM_011520816.2 | Intron | XP_011519118.1 | ||||
XM_017019797.1 | Intron | XP_016875286.1 |