Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606444 MIM: 612988 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CREBZF PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CREBZF - CREB/ATF bZIP transcription factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039618.2 | 4253 | UTR 3 | NP_001034707.1 | |||
XM_006718642.3 | 4253 | UTR 3 | XP_006718705.1 | |||
XM_011545195.2 | 4253 | UTR 3 | XP_011543497.1 | |||
XM_017018086.1 | 4253 | UTR 3 | XP_016873575.1 | |||
XM_017018087.1 | 4253 | UTR 3 | XP_016873576.1 | |||
XM_017018088.1 | 4253 | UTR 3 | XP_016873577.1 | |||
XM_017018089.1 | 4253 | UTR 3 | XP_016873578.1 | |||
XM_017018090.1 | 4253 | UTR 3 | XP_016873579.1 | |||
XM_017018091.1 | 4253 | UTR 3 | XP_016873580.1 | |||
XM_017018092.1 | 4253 | UTR 3 | XP_016873581.1 |
TMEM126A - transmembrane protein 126A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |