Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
74 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616751 MIM: 615111 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEPT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CEPT1 - choline/ethanolamine phosphotransferase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DENND2D - DENN domain containing 2D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271833.1 | 1435 | Silent Mutation | GAA,GAG | E,E 394 | NP_001258762.1 | |
NM_024901.4 | 1435 | Silent Mutation | GAA,GAG | E,E 397 | NP_079177.2 | |
XM_006710921.2 | 1435 | Silent Mutation | GAA,GAG | E,E 411 | XP_006710984.2 | |
XM_006710922.1 | 1435 | Silent Mutation | GAA,GAG | E,E 341 | XP_006710985.1 | |
XM_006710923.1 | 1435 | Silent Mutation | GAA,GAG | E,E 341 | XP_006710986.1 | |
XM_006710924.1 | 1435 | Silent Mutation | GAA,GAG | E,E 341 | XP_006710987.1 | |
XM_011542187.1 | 1435 | Silent Mutation | GAA,GAG | E,E 385 | XP_011540489.1 | |
XM_011542189.1 | 1435 | Silent Mutation | GAA,GAG | E,E 341 | XP_011540491.1 | |
XM_011542190.2 | 1435 | Silent Mutation | GAA,GAG | E,E 385 | XP_011540492.1 | |
XM_017002386.1 | 1435 | Silent Mutation | GAA,GAG | E,E 217 | XP_016857875.1 | |
XM_017002387.1 | 1435 | Silent Mutation | GAA,GAG | E,E 217 | XP_016857876.1 | |
XM_017002388.1 | 1435 | Silent Mutation | GAA,GAG | E,E 217 | XP_016857877.1 | |
XM_017002389.1 | 1435 | Intron | XP_016857878.1 |