Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGCCATATCTACAGTTAAGCCAT[G/T]TATTGCCTCTTGCAACTCTGGTAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120355 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MMP27 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MMP27 - matrix metallopeptidase 27 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MMP8 - matrix metallopeptidase 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304441.1 | 1558 | Missense Mutation | AAA,ACA | K,T 437 | NP_001291370.1 | |
NM_001304442.1 | 1558 | Missense Mutation | AAA,ACA | K,T 437 | NP_001291371.1 | |
NM_002424.2 | 1558 | Missense Mutation | AAA,ACA | K,T 460 | NP_002415.1 | |
XM_011542834.2 | 1558 | Missense Mutation | AAA,ACA | K,T 469 | XP_011541136.1 | |
XM_011542835.2 | 1558 | Missense Mutation | AAA,ACA | K,T 437 | XP_011541137.1 | |
XM_011542836.2 | 1558 | UTR 3 | XP_011541138.1 | |||
XM_017017771.1 | 1558 | Missense Mutation | AAA,ACA | K,T 437 | XP_016873260.1 |
Set Membership: |
Validated |