Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612927 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AVL9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AVL9 - AVL9 cell migration associated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015060.2 | 4406 | UTR 3 | NP_055875.1 | |||
XM_005249668.2 | 4406 | UTR 3 | XP_005249725.1 | |||
XM_005249669.2 | 4406 | UTR 3 | XP_005249726.1 | |||
XM_005249670.2 | 4406 | UTR 3 | XP_005249727.1 | |||
XM_005249671.4 | 4406 | Intron | XP_005249728.1 | |||
XM_011515227.2 | 4406 | UTR 3 | XP_011513529.1 | |||
XM_017011891.1 | 4406 | UTR 3 | XP_016867380.1 | |||
XM_017011892.1 | 4406 | UTR 3 | XP_016867381.1 |
DPY19L1P1 - DPY19L1 pseudogene 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |