Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGGCCTTCCTCTAGGGCACCGCA[C/T]GCCCCTTCCCCCTCGCCCACGTCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616159 MIM: 612275 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DHRS11 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DHRS11 - dehydrogenase/reductase 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024308.3 | Intron | NP_077284.2 | ||||
XM_005257658.2 | Intron | XP_005257715.1 | ||||
XM_011525233.2 | Intron | XP_011523535.1 |
GGNBP2 - gametogenetin binding protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRM1 - mitochondrial rRNA methyltransferase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |