Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCAAAAGTTATGATTATGGTCAT[C/G]TAAGCCACAGGATCTGTTTTCTTAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 179550 | ||||||||||||||||||||
Literature Links: |
RALA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RALA - RALA Ras like proto-oncogene A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005402.3 | Intron | NP_005393.2 | ||||
XM_006715762.2 | Intron | XP_006715825.1 | ||||
XM_011515466.1 | Intron | XP_011513768.1 | ||||
XM_011515467.1 | Intron | XP_011513769.1 |