Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608595 MIM: 608596 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NPSR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NPSR1 - neuropeptide S receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300933.1 | 72 | UTR 5 | NP_001287862.1 | |||
NM_001300934.1 | 72 | UTR 5 | NP_001287863.1 | |||
NM_001300935.1 | 72 | UTR 5 | NP_001287864.1 | |||
NM_207172.1 | 72 | UTR 5 | NP_997055.1 | |||
NM_207173.1 | 72 | UTR 5 | NP_997056.1 |
NPSR1-AS1 - NPSR1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |