Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604842 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC22A3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SLC22A3 - solute carrier family 22 member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021977.3 | 402 | Silent Mutation | CGC,CGT | R,R 120 | NP_068812.1 | |
XM_005267106.4 | 402 | Intron | XP_005267163.1 | |||
XM_005267107.3 | 402 | Silent Mutation | CGC,CGT | R,R 120 | XP_005267164.1 | |
XM_011536075.2 | 402 | Intron | XP_011534377.1 | |||
XM_011536076.2 | 402 | Intron | XP_011534378.1 | |||
XM_017011203.1 | 402 | Intron | XP_016866692.1 |
Set Membership: |
DME Validated Inventoried |