Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTCTCAACAGTTCAGGACTTTGG[A/C]GGCAAAGAGAGACGGGGCTGTGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||
Literature Links: |
FOXJ2 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
FOXJ2 - forkhead box J2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018416.2 | 568 | UTR 5 | NP_060886.1 | |||
XM_011520760.2 | 568 | UTR 5 | XP_011519062.1 | |||
XM_011520761.2 | 568 | UTR 5 | XP_011519063.1 | |||
XM_017019640.1 | 568 | Intron | XP_016875129.1 |
Set Membership: |
HapMap |