Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
30 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605226 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RERE PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RERE - arginine-glutamic acid dipeptide repeats | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042681.1 | Intron | NP_001036146.1 | ||||
NM_001042682.1 | Intron | NP_001036147.1 | ||||
NM_012102.3 | Intron | NP_036234.3 | ||||
XM_005263464.2 | Intron | XP_005263521.1 | ||||
XM_005263466.1 | Intron | XP_005263523.1 | ||||
XM_011541510.1 | Intron | XP_011539812.1 | ||||
XM_011541511.1 | Intron | XP_011539813.1 | ||||
XM_017001358.1 | Intron | XP_016856847.1 | ||||
XM_017001359.1 | Intron | XP_016856848.1 |