Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601041 MIM: 612399 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TLE2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TLE2 - transducin like enhancer of split 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144761.1 | Intron | NP_001138233.1 | ||||
NM_001144762.1 | Intron | NP_001138234.1 | ||||
NM_001300846.1 | Intron | NP_001287775.1 | ||||
NM_003260.4 | Intron | NP_003251.2 | ||||
XM_006722864.2 | Intron | XP_006722927.1 | ||||
XM_011528230.1 | Intron | XP_011526532.1 |
TLE6 - transducin like enhancer of split 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |