Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATATGAAAACAAATTTGTATCTCTT[C/T]CTTTTCACATGATTTATGTGGCAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
38 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612176 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYSM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MYSM1 - Myb like, SWIRM and MPN domains 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001085487.2 | 2644 | UTR 3 | NP_001078956.1 | |||
XM_006710314.3 | 2644 | UTR 3 | XP_006710377.1 | |||
XM_011540573.2 | 2644 | UTR 3 | XP_011538875.1 | |||
XM_011540574.2 | 2644 | UTR 3 | XP_011538876.1 | |||
XM_017000195.1 | 2644 | UTR 3 | XP_016855684.1 | |||
XM_017000196.1 | 2644 | UTR 3 | XP_016855685.1 | |||
XM_017000197.1 | 2644 | UTR 3 | XP_016855686.1 |