Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTATCTTCACCTGCCTGAATGGA[A/G]CAGCCCTCTCTCACTCGGTTCTGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BEND2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BEND2 - BEN domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184767.1 | Intron | NP_001171696.1 | ||||
NM_153346.4 | Intron | NP_699177.2 | ||||
XM_011545447.1 | Intron | XP_011543749.1 | ||||
XM_017029266.1 | Intron | XP_016884755.1 | ||||
XM_017029267.1 | Intron | XP_016884756.1 |