Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTGAGCTTTACCGTCCAGTCTTC[A/G]TCACTGTGGTTTTGTAGTGATTTCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176640 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRNP PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PRNP - prion protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000311.3 | Intron | NP_000302.1 | ||||
NM_001080121.1 | Intron | NP_001073590.1 | ||||
NM_001080122.1 | Intron | NP_001073591.1 | ||||
NM_001080123.1 | Intron | NP_001073592.1 | ||||
NM_001271561.1 | Intron | NP_001258490.1 | ||||
NM_183079.2 | Intron | NP_898902.1 |