Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 116899 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDKN1A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDKN1A - cyclin dependent kinase inhibitor 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000389.4 | 150 | Intron | NP_000380.1 | |||
NM_001220777.1 | 150 | Intron | NP_001207706.1 | |||
NM_001220778.1 | 150 | Intron | NP_001207707.1 | |||
NM_001291549.1 | 150 | UTR 5 | NP_001278478.1 | |||
NM_078467.2 | 150 | Intron | NP_510867.1 |
PANDAR - promoter of CDKN1A antisense DNA damage activated RNA | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |