Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGAGAAGGGAAAAGGACTGGGTTT[C/G]TGGATGCTGAGCTCAGCAGCCTCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
FAM163B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM163B - family with sequence similarity 163 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080515.2 | Intron | NP_001073984.1 | ||||
XM_005272203.4 | Intron | XP_005272260.1 | ||||
XM_005272204.4 | Intron | XP_005272261.1 | ||||
XM_006717233.3 | Intron | XP_006717296.1 | ||||
XM_011518912.2 | Intron | XP_011517214.1 | ||||
XM_011518913.2 | Intron | XP_011517215.1 | ||||
XM_017015017.1 | Intron | XP_016870506.1 | ||||
XM_017015018.1 | Intron | XP_016870507.1 |