Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613599 MIM: 138550 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABHD12 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ABHD12 - abhydrolase domain containing 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042472.2 | Intron | NP_001035937.1 | ||||
NM_015600.4 | Intron | NP_056415.1 | ||||
XM_005260698.1 | Intron | XP_005260755.1 | ||||
XM_011529214.2 | Intron | XP_011527516.1 | ||||
XM_011529215.1 | Intron | XP_011527517.1 | ||||
XM_011529218.1 | Intron | XP_011527520.1 | ||||
XM_017027796.1 | Intron | XP_016883285.1 | ||||
XM_017027797.1 | Intron | XP_016883286.1 | ||||
XM_017027798.1 | Intron | XP_016883287.1 |
PYGB - phosphorylase, glycogen; brain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002862.3 | Intron | NP_002853.2 |
Set Membership: |
HapMap |