Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602004 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPDPF PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PPDPF - pancreatic progenitor cell differentiation and proliferation factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PTK6 - protein tyrosine kinase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256358.1 | Intron | NP_001243287.1 | ||||
NM_005975.3 | Intron | NP_005966.1 | ||||
XM_017027982.1 | Intron | XP_016883471.1 |
SRMS - src-related kinase lacking C-terminal regulatory tyrosine and N-terminal myristylation sites | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |