Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTACATATTTTATATTTATGAAA[G/T]GGCACTGGGAGAGGGTTGTTACTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612994 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAB28 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RAB28 - RAB28, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017979.2 | Intron | NP_001017979.1 | ||||
NM_001159601.1 | Intron | NP_001153073.1 | ||||
NM_004249.3 | Intron | NP_004240.2 | ||||
XM_005248218.4 | Intron | XP_005248275.1 | ||||
XM_011513911.2 | Intron | XP_011512213.1 | ||||
XM_011513912.2 | Intron | XP_011512214.1 | ||||
XM_017008850.1 | Intron | XP_016864339.1 |