Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACATTATGAAAAAGTTCATAATC[C/G]ACTATGATGATACAATAATTATGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
42 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MROH9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MROH9 - maestro heat like repeat family member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163629.1 | Intron | NP_001157101.1 | ||||
NM_025063.2 | Intron | NP_079339.2 | ||||
XM_011510005.2 | Intron | XP_011508307.1 | ||||
XM_011510006.2 | Intron | XP_011508308.1 | ||||
XM_011510007.2 | Intron | XP_011508309.1 |