Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
44 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SUCO PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SUCO - SUN domain containing ossification factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282750.1 | 303 | UTR 5 | NP_001269679.1 | |||
NM_001282751.1 | 303 | UTR 5 | NP_001269680.1 | |||
NM_014283.4 | 303 | UTR 5 | NP_055098.1 | |||
NM_016227.3 | 303 | Silent Mutation | ATC,ATT | I,I 163 | NP_057311.3 | |
XM_006711374.2 | 303 | UTR 5 | XP_006711437.1 | |||
XM_006711375.2 | 303 | UTR 5 | XP_006711438.1 | |||
XM_006711376.1 | 303 | UTR 5 | XP_006711439.1 |
Set Membership: |
JSNP Validated |