Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614855 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TBC1D14 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
TBC1D14 - TBC1 domain family member 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001113361.1 | Intron | NP_001106832.1 | ||||
NM_001113363.1 | Intron | NP_001106834.1 | ||||
NM_001286805.1 | Intron | NP_001273734.1 | ||||
NM_020773.2 | Intron | NP_065824.2 | ||||
XM_005247986.2 | Intron | XP_005248043.1 | ||||
XM_006713895.2 | Intron | XP_006713958.1 | ||||
XM_011513507.2 | Intron | XP_011511809.1 | ||||
XM_011513508.1 | Intron | XP_011511810.1 | ||||
XM_011513513.2 | Intron | XP_011511815.1 | ||||
XM_017008477.1 | Intron | XP_016863966.1 | ||||
XM_017008478.1 | Intron | XP_016863967.1 | ||||
XM_017008479.1 | Intron | XP_016863968.1 | ||||
XM_017008480.1 | Intron | XP_016863969.1 |
Set Membership: |
HapMap Validated |