Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAATACTCTCTCTGCATCTATCGA[A/G]ATGATCATATGGTTTTTGTACTTCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605093 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SH2B3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SH2B3 - SH2B adaptor protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291424.1 | Intron | NP_001278353.1 | ||||
NM_005475.2 | Intron | NP_005466.1 | ||||
XM_005253818.4 | Intron | XP_005253875.1 | ||||
XM_005253819.4 | Intron | XP_005253876.1 | ||||
XM_006719180.3 | Intron | XP_006719243.1 | ||||
XM_011537719.2 | Intron | XP_011536021.1 | ||||
XM_011537720.2 | Intron | XP_011536022.1 | ||||
XM_011537721.2 | Intron | XP_011536023.1 |
Set Membership: |
HapMap |