Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGGGGTGGGGGTGGGGGAGGAGG[A/G]AGAGACATAAGCCTGAGTTGGAGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 186946 MIM: 600230 MIM: 601140 MIM: 601398 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FKBP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FKBP2 - FK506 binding protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC105369340 - uncharacterized LOC105369340 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLCB3 - phospholipase C beta 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPP1R14B - protein phosphatase 1 regulatory inhibitor subunit 14B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138689.2 | Intron | NP_619634.1 |
VEGFB - vascular endothelial growth factor B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |