Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TTC7B PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
TTC7B - tetratricopeptide repeat domain 7B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010854.1 | Intron | NP_001010854.1 | ||||
NM_001320421.1 | Intron | NP_001307350.1 | ||||
XM_005267367.1 | Intron | XP_005267424.1 | ||||
XM_011536497.1 | Intron | XP_011534799.1 | ||||
XM_017021044.1 | Intron | XP_016876533.1 | ||||
XM_017021045.1 | Intron | XP_016876534.1 | ||||
XM_017021046.1 | Intron | XP_016876535.1 | ||||
XM_017021047.1 | Intron | XP_016876536.1 |