Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACCAAATCATGGTGCACTAACAG[G/T]CATACTGTAGGTGCCCAGGGAAAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC85C PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CCDC85C - coiled-coil domain containing 85C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HHIPL1 - HHIP like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_006720277.3 | Intron | XP_006720340.1 | ||||
XM_011537236.2 | Intron | XP_011535538.1 | ||||
XM_011537237.2 | Intron | XP_011535539.1 | ||||
XM_017021707.1 | Intron | XP_016877196.1 | ||||
XM_017021708.1 | Intron | XP_016877197.1 |
Set Membership: |
Validated |