Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCACCTCTAGTAAACTTTCTCCC[C/T]CAGTTTTCTATAATATATTGGATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608132 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TTC8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TTC8 - tetratricopeptide repeat domain 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288781.1 | Intron | NP_001275710.1 | ||||
NM_001288782.1 | Intron | NP_001275711.1 | ||||
NM_001288783.1 | Intron | NP_001275712.1 | ||||
NM_144596.3 | Intron | NP_653197.2 | ||||
NM_198309.3 | Intron | NP_938051.1 | ||||
NM_198310.3 | Intron | NP_938052.1 | ||||
XM_006720035.1 | Intron | XP_006720098.1 | ||||
XM_006720037.2 | Intron | XP_006720100.1 | ||||
XM_011536432.1 | Intron | XP_011534734.1 | ||||
XM_011536433.1 | Intron | XP_011534735.1 | ||||
XM_011536434.1 | Intron | XP_011534736.1 | ||||
XM_011536435.2 | Intron | XP_011534737.1 | ||||
XM_017020977.1 | Intron | XP_016876466.1 | ||||
XM_017020978.1 | Intron | XP_016876467.1 |