Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C14orf159 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
C14orf159 - chromosome 14 open reading frame 159 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001102366.2 | Intron | NP_001095836.1 | ||||
NM_001102367.2 | Intron | NP_001095837.1 | ||||
NM_001102368.2 | Intron | NP_001095838.1 | ||||
NM_001102369.2 | Intron | NP_001095839.1 | ||||
NM_001286470.1 | Intron | NP_001273399.1 | ||||
NM_001286471.1 | Intron | NP_001273400.1 | ||||
NM_001286472.1 | Intron | NP_001273401.1 | ||||
NM_001286473.1 | Intron | NP_001273402.1 | ||||
NM_024952.7 | Intron | NP_079228.4 |
SNORA11B - small nucleolar RNA, H/ACA box 11B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |