Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C14orf159 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C14orf159 - chromosome 14 open reading frame 159 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001102366.2 | Intron | NP_001095836.1 | ||||
NM_001102367.2 | Intron | NP_001095837.1 | ||||
NM_001102368.2 | Intron | NP_001095838.1 | ||||
NM_001102369.2 | Intron | NP_001095839.1 | ||||
NM_001286470.1 | Intron | NP_001273399.1 | ||||
NM_001286471.1 | Intron | NP_001273400.1 | ||||
NM_001286472.1 | Intron | NP_001273401.1 | ||||
NM_001286473.1 | Intron | NP_001273402.1 | ||||
NM_024952.7 | Intron | NP_079228.4 |