Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611520 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
POLDIP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
POLDIP3 - DNA polymerase delta interacting protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278657.1 | 1660 | UTR 3 | NP_001265586.1 | |||
NM_032311.4 | 1660 | UTR 3 | NP_115687.2 | |||
NM_178136.2 | 1660 | UTR 3 | NP_835237.1 | |||
XM_011530457.1 | 1660 | UTR 3 | XP_011528759.1 | |||
XM_017029010.1 | 1660 | UTR 3 | XP_016884499.1 | |||
XM_017029011.1 | 1660 | Intron | XP_016884500.1 |
RRP7BP - ribosomal RNA processing 7 homolog B, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |