Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613213 MIM: 611520 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYB5R3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CYB5R3 - cytochrome b5 reductase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000398.6 | 1193 | UTR 3 | NP_000389.1 | |||
NM_001129819.2 | 1193 | UTR 3 | NP_001123291.1 | |||
NM_001171660.1 | 1193 | UTR 3 | NP_001165131.1 | |||
NM_001171661.1 | 1193 | UTR 3 | NP_001165132.1 | |||
NM_007326.4 | 1193 | UTR 3 | NP_015565.1 |
POLDIP3 - DNA polymerase delta interacting protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNU12 - RNA, U12 small nuclear | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |