Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCAGCTGGGCAGTGCAGTGCGCC[C/T]GGGAAAGGACTCTGCCTCTGGAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613213 MIM: 611520 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYB5R3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CYB5R3 - cytochrome b5 reductase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000398.6 | Intron | NP_000389.1 | ||||
NM_001129819.2 | Intron | NP_001123291.1 | ||||
NM_001171660.1 | Intron | NP_001165131.1 | ||||
NM_001171661.1 | Intron | NP_001165132.1 | ||||
NM_007326.4 | Intron | NP_015565.1 |
POLDIP3 - DNA polymerase delta interacting protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNU12 - RNA, U12 small nuclear | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |